Learn More
Waters Corp WATERS CORPORATION Guard Cartridge, MS C18, 5 μm, 30 x 10 mm
Catalog No. 502391645 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge features MS C18 chemistry with a 5 μm particle size and 30 x 10 mm dimensions, suitable for preparative column applications. It requires a compatible 30 x 10 mm cartridge holder.
  • Designed to be compatible with mass spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Compatible with pH range 1–12
Catalog No. 50-239-1645 Supplier Waters Corp Supplier No. 186006903
This guard cartridge features MS C18 chemistry with a 5 μm particle size and 30 x 10 mm dimensions, suitable for preparative column applications. It requires a compatible 30 x 10 mm cartridge holder.
  • Designed to be compatible with mass spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Compatible with pH range 1–12