Learn More
Waters Corp Guard Cartridge, C18, 125Å, 10 μm, 30 mm x 10 mm
Catalog No. 502391444 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge, part of the column line, utilizes Hybrid Particle Technology for isolation and purification applications. The patented Hybrid Particle Technology is specifically engineered for prep, enabling routine operation at pH 1 to 12.
  • Uses patented Hybrid Particle Technology.
  • Designed for isolation and purification applications.
  • Operates routinely at pH 1 to 12.
  • Allows loading up to 60X more onto the column.
  • Provides greater capacity, leading to fewer injections for the same amount of pure material.
  • Can produce more material with the same number of injections.
  • Enables the use of shorter columns in discovery applications, producing material in half the time.
Catalog No. 50-239-1444 Supplier Waters Corp Supplier No. 186006902
This guard cartridge, part of the column line, utilizes Hybrid Particle Technology for isolation and purification applications. The patented Hybrid Particle Technology is specifically engineered for prep, enabling routine operation at pH 1 to 12.
  • Uses patented Hybrid Particle Technology.
  • Designed for isolation and purification applications.
  • Operates routinely at pH 1 to 12.
  • Allows loading up to 60X more onto the column.
  • Provides greater capacity, leading to fewer injections for the same amount of pure material.
  • Can produce more material with the same number of injections.
  • Enables the use of shorter columns in discovery applications, producing material in half the time.