Learn More
Waters Corp XTerra VanGuard Cartridge, C18, 125Å, 2.5 μm, 2.1 mm x 5 mm
Catalog No. 502390557 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase. They are optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Extends analytical column lifetime
  • Enhances analytical column performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns
Catalog No. 50-239-0557 Supplier Waters Corp Supplier No. 186007887
XTerra MS C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase. They are optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Extends analytical column lifetime
  • Enhances analytical column performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns