Learn More
Waters Corp VanGuard Cartridge, C18, 2.5 μm, 3.9 mm × 5 mm
Catalog No. 502392156 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges are designed to extend analytical column lifetime and performance. They achieve this by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns that contain 2.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 3.9 mm and 4.6 mm I.D. MS C18 analytical columns containing 2.5 μm sorbent particles.
Catalog No. 50-239-2156 Supplier Waters Corp Supplier No. 186007889
VanGuard Cartridges are designed to extend analytical column lifetime and performance. They achieve this by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns that contain 2.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 3.9 mm and 4.6 mm I.D. MS C18 analytical columns containing 2.5 μm sorbent particles.