Learn More
Waters Corp VanGuard Cartridge, C18, 5 μm, 3.9 mm x 5 mm
Catalog No. 502392643 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 VanGuard Cartridges extend analytical column lifespan by removing particulate mobile phase contamination. Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent, they install easily. Cartridges shield columns, improve performance by filtering impurities, and maintain UPLC, UHPLC, and HPLC separation efficiency.
  • Hybrid particle technology: silica efficiency with polymer stability for wide pH (1–12) operation.
  • Ensures excellent peak shapes for bases and high mechanical strength.
  • MS compatibility provides sharp peaks, good sensitivity, and ultra-low bleed.
  • QC tested with peptide maps for consistent performance and extended column life.
  • Sustains high pressures in UPLC system applications; allows easy scale-up.
Catalog No. 50-239-2643 Supplier Waters Corp Supplier No. 186007899
XTerra MS C18 VanGuard Cartridges extend analytical column lifespan by removing particulate mobile phase contamination. Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent, they install easily. Cartridges shield columns, improve performance by filtering impurities, and maintain UPLC, UHPLC, and HPLC separation efficiency.
  • Hybrid particle technology: silica efficiency with polymer stability for wide pH (1–12) operation.
  • Ensures excellent peak shapes for bases and high mechanical strength.
  • MS compatibility provides sharp peaks, good sensitivity, and ultra-low bleed.
  • QC tested with peptide maps for consistent performance and extended column life.
  • Sustains high pressures in UPLC system applications; allows easy scale-up.