Learn More
Waters Corp VanGuard Cartridge, MS C18, 125Å, 3.5 μm, 2.1 mm X 5 mm
Catalog No. 502392637 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge extends analytical column lifespan and performance by filtering particulate contamination from the mobile phase. Optimized for 2.1 mm and 3.0 mm I.D. columns with 3.5 μm sorbent particles, it is easy to set up, defending against contaminants and boosting analytical performance.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contamination.
  • Optimized for 2.1 mm and 3.0 mm I.D. columns with 3.5 μm sorbent particles.
  • Easy to operate and set up.
  • Maintains UPLC, UHPLC, and HPLC separation performance.
  • Designed for high-pressure ACQUITY UPLC System applications.
  • Ensures consistent performance via peptide map QC.
  • Single component for direct fitting to UPLC Column intake.
Catalog No. 50-239-2637 Supplier Waters Corp Supplier No. 186007892
This cartridge extends analytical column lifespan and performance by filtering particulate contamination from the mobile phase. Optimized for 2.1 mm and 3.0 mm I.D. columns with 3.5 μm sorbent particles, it is easy to set up, defending against contaminants and boosting analytical performance.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contamination.
  • Optimized for 2.1 mm and 3.0 mm I.D. columns with 3.5 μm sorbent particles.
  • Easy to operate and set up.
  • Maintains UPLC, UHPLC, and HPLC separation performance.
  • Designed for high-pressure ACQUITY UPLC System applications.
  • Ensures consistent performance via peptide map QC.
  • Single component for direct fitting to UPLC Column intake.