Learn More
Waters Corp HPLC Cartridge, Hybrid, 125 Å, 3.5 μm
Supplier:  Waters Corp 186007894
Encompass_Preferred
This HPLC cartridge is a hybrid, spherical particle column designed for robust chromatographic separations. It features a low silanol activity and is end-capped, ensuring reliable performance across a wide pH range.
  • Hybrid material construction
  • USP Class L1
  • pH range of 1 to 12
  • Low silanol activity for improved peak shape
  • End-capped to minimize secondary interactions
  • 15.5% carbon load
  • Reversed phase (RP) mode
  • 3.9 mm inner diameter
  • 5 mm length
  • 3.5 μm particle size
  • 125 Å pore size
Catalog No. 50-239-2527