Learn More
Waters Corp Waters Corporation XTerra MS C8 Column, 3.5 μm, 2.1 x 100 mm, 125 Å
Supplier:  Waters Corp 186000405
Encompass_Preferred
These XTerra MS C8 Columns are designed for compatibility with Mass Spectrometry applications, offering sharp peaks, good sensitivity, and large peak capacities.
  • Feature trifunctional bonding chemistries.
  • Deliver long lifetimes over a wide pH range (1–12).
  • Provide high throughput with excellent resolution.
  • Offer ultra-low bleed.
  • Ideally suited for Mass Spec applications.
Catalog No. 50-780-224