Learn More
Waters Corp Waters HPLC Column, 3.9 x 100 mm, 3.5 μm, MS C8
Catalog No. 50780246 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns are hybrid particle (silica co–polymer) stationary phases designed for Mass Spectrometry applications. They combine high efficiency with an expanded pH range (1–12), offering sharp peaks, good sensitivity, and large peak capacities. Featuring trifunctional bonding, these columns provide long lifetimes, high throughput, excellent resolution, and ultra–low bleed.
  • Hybrid particle (silica co–polymer)
  • High mechanical strength
  • High efficiency
  • Excellent peak shape for bases
  • Wider pH range (1–12)
  • Compatible with Mass Spectrometry applications
  • Sharp peaks
  • Good sensitivity
  • Large peak capacities
  • Trifunctional bonding
  • Long lifetimes
  • High throughput with excellent resolution
  • Ultra–low bleed
Catalog No. 50-780-246 Supplier Waters Corp Supplier No. 186000427
XTerra HPLC Columns are hybrid particle (silica co–polymer) stationary phases designed for Mass Spectrometry applications. They combine high efficiency with an expanded pH range (1–12), offering sharp peaks, good sensitivity, and large peak capacities. Featuring trifunctional bonding, these columns provide long lifetimes, high throughput, excellent resolution, and ultra–low bleed.
  • Hybrid particle (silica co–polymer)
  • High mechanical strength
  • High efficiency
  • Excellent peak shape for bases
  • Wider pH range (1–12)
  • Compatible with Mass Spectrometry applications
  • Sharp peaks
  • Good sensitivity
  • Large peak capacities
  • Trifunctional bonding
  • Long lifetimes
  • High throughput with excellent resolution
  • Ultra–low bleed