Learn More
Waters Corp Waters XTerra MS C8 Column, 3.5 μm Particle, 3.0 x 100 mm
Catalog No. 50780238 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns combine the high efficiency separations of a silica particle with the expanded pH range of a polymer particle. This results in a rugged material with high mechanical strength, high efficiency, excellent peak shape for bases, and a wider pH range (1–12) than silica–based particles. The particle shape is spherical and the pore size is 125 Å.
  • Designed to be compatible with Mass Spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Features sharpest peaks
  • Trifunctional bonding
  • Longest lifetimes
  • Ideally suited for Mass Spec
Catalog No. 50-780-238 Supplier Waters Corp Supplier No. 186000419
XTerra HPLC Columns combine the high efficiency separations of a silica particle with the expanded pH range of a polymer particle. This results in a rugged material with high mechanical strength, high efficiency, excellent peak shape for bases, and a wider pH range (1–12) than silica–based particles. The particle shape is spherical and the pore size is 125 Å.
  • Designed to be compatible with Mass Spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Features sharpest peaks
  • Trifunctional bonding
  • Longest lifetimes
  • Ideally suited for Mass Spec