Learn More
Waters Corp Waters Corporation XTerra MS C8 Column, 125 Å, 3.5 μm, 3.0 × 30 mm
Supplier:  Waters Corp 186000413
Encompass_Preferred
The XTerra MS C8 Column is designed for Mass Spectrometry applications, providing sharp peaks, good sensitivity, and large peak capacities. It features trifunctional bonding chemistries for long lifetimes across a wide pH range and offers high throughput with excellent resolution and ultra-low bleed.
  • Compatible with mass spectrometry applications.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range.
  • Provides high throughput with excellent resolution and ultra-low bleed.
  • Offers sharpest peaks.
  • pH range 1 to 12.
  • Ideally suited for mass spec.
Catalog No. 50-780-232