Learn More
Waters Corp Waters XTerra MS C8 Column, 3.5 μm, 4.6 mm × 250 mm, 125 Å
Supplier:  Waters Corp 186001471
Encompass_Preferred
The XTerra MS C8 column is a general-purpose, hybrid-based, reversed-phase C8 column. It is specifically designed for compatibility with mass spectrometry applications, offering superior pH stability compared to silica-based columns.
  • Designed for compatibility with mass spectrometry applications.
  • Provides superior pH stability compared to silica-based columns.
  • Delivers sharp peaks, good sensitivity, and large peak capacities.
  • Utilizes trifunctional bonding chemistries for long lifetimes over a wide pH range.
  • Offers high throughput with excellent resolution and ultra-low bleed.
Catalog No. 50-780-700