Learn More
Waters Corp XTerra MS C8 Column, 125Å, 3.5 µm, 4.6 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186000437
Encompass_Preferred
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns that are designed to be compatible with mass spectrometry applications. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-256