Learn More
Waters Corp Waters XTerra MS C8 Column, 5 μm, 2.1 x 100 mm
Supplier:  Waters Corp 186000451
Encompass_Preferred
This XTerra MS C8 Column is an analytical column with a 2.1 x 100 mm dimension and 5 μm particle size, specifically engineered for Mass Spectrometry applications. Its trifunctional bonding chemistries and hybrid particle technology ensure durability and stable performance across a broad pH range of 1 to 12. The column delivers sharp peaks, excellent resolution, and high throughput, making it ideal for demanding MS analyses.
  • First-generation hybrid particle for high efficiency separations and expanded pH range.
  • Designed for mass spectrometry (MS) applications, providing sharp peaks and good sensitivity.
  • Trifunctional bonding chemistries ensure long lifetimes over a wide pH range (1 to 12).
  • Offers high throughput, excellent resolution, and ultra-low bleed.
  • Rugged material with high mechanical strength and excellent peak shape for bases.
Catalog No. 50-780-268