Learn More
Waters Corp Waters Column, C8, 5 μm, 2.1 mm X 150 mm, 125 Å
Catalog No. 50780272 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general-purpose, hybrid-based, reversed-phase C8 columns are engineered for compatibility with mass spectrometry applications. They offer superior pH stability compared to silica-based columns.
  • C8 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • MS C8 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 125 Å pore size
  • Surface area of 175
  • Compatible with HPLC systems
  • USP classification L7
  • 2.1 mm inner diameter
  • 150 mm length
  • 12% carbon load
  • 1 unit per package
Catalog No. 50-780-272 Supplier Waters Corp Supplier No. 186000455
These general-purpose, hybrid-based, reversed-phase C8 columns are engineered for compatibility with mass spectrometry applications. They offer superior pH stability compared to silica-based columns.
  • C8 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • MS C8 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 125 Å pore size
  • Surface area of 175
  • Compatible with HPLC systems
  • USP classification L7
  • 2.1 mm inner diameter
  • 150 mm length
  • 12% carbon load
  • 1 unit per package