Learn More
Waters Corp XTerra MS C8 Column, 125Å, 5 μm, 4.6 mm X 250 mm
Catalog No. 50780312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C8 columns are general-purpose, hybrid-based, reversed-phase C8 columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • Compatible with mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.
Catalog No. 50-780-312 Supplier Waters Corp Supplier No. 186000495
Add to Cart
Add to Cart
XTerra MS C8 columns are general-purpose, hybrid-based, reversed-phase C8 columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • Compatible with mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.