Learn More
Waters Corp WATERS Column, C8, 5 μm, 4.6 mm X 150 mm, 125 Å
Catalog No. 50819080 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C8 columns are general-purpose, hybrid-based, reversed-phase C8 columns compatible with mass spectrometry applications. They provide superior pH stability compared to silica-based columns, delivering crisp peak forms, high sensitivity, and large peak capacities. Utilizing tri-functional bonding chemistry, these columns offer extended life over a wide pH range, high throughput, excellent resolution, and minimal column bleed.
  • Compatible with mass spectrometry applications.
  • Uses Hybrid Particle Technology with inorganic silica and organic organosiloxane components.
  • Offers separation efficiency and improved pH stability.
  • Each column is thoroughly examined, and includes Certificates of Batch Analysis and Column Efficiency.
  • Maximum pressure capability: 6000 psi (415 Bar).
  • Working pH range: 1 to 12.
Catalog No. 50-819-080 Supplier Waters Corp Supplier No. 186000491
Add to Cart
Add to Cart
XTerra MS C8 columns are general-purpose, hybrid-based, reversed-phase C8 columns compatible with mass spectrometry applications. They provide superior pH stability compared to silica-based columns, delivering crisp peak forms, high sensitivity, and large peak capacities. Utilizing tri-functional bonding chemistry, these columns offer extended life over a wide pH range, high throughput, excellent resolution, and minimal column bleed.
  • Compatible with mass spectrometry applications.
  • Uses Hybrid Particle Technology with inorganic silica and organic organosiloxane components.
  • Offers separation efficiency and improved pH stability.
  • Each column is thoroughly examined, and includes Certificates of Batch Analysis and Column Efficiency.
  • Maximum pressure capability: 6000 psi (415 Bar).
  • Working pH range: 1 to 12.