Learn More
Waters Corp Waters XTerra MS C8 Sentry Guard Cartridge, 5 μm, 3.9 mm x 20 mm
Catalog No. 50780450 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns designed for compatibility with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • General purpose, hybrid-based, reversed-phase C8 columns.
  • Designed to be compatible with mass spectrometry applications.
  • Provides superior pH stability compared to silica-based columns.
Catalog No. 50-780-450 Supplier Waters Corp Supplier No. 186000661
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns designed for compatibility with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • General purpose, hybrid-based, reversed-phase C8 columns.
  • Designed to be compatible with mass spectrometry applications.
  • Provides superior pH stability compared to silica-based columns.