Learn More
Waters Corp Waters HPLC Column, MS C8, 10 x 250 mm, 10 μm
Catalog No. 502392449 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HPLC columns are the first commercially available hybrid particle (silica co-polymer) stationary phase, providing the efficiency of silica with the stability of polymers. This C8 column is designed for Mass Spectrometry applications, offering sharp peaks, good sensitivity, and large peak capacities.
  • Designed with trifunctional bonding chemistries for long lifetimes over a wide pH range.
  • Provides high throughput, excellent resolution, and ultra-low bleed.
  • Features homogenous particle coverage.
  • Contains one-third less silanols for superior peak shape.
  • Operates within a pH range of 1–12.
  • Constructed from hybrid particle (silica co-polymer) material.
  • Utilizes methylene bridged particles with 12% carbon load.
Catalog No. 50-239-2449 Supplier Waters Corp Supplier No. 186008134
Add to Cart
Add to Cart
These HPLC columns are the first commercially available hybrid particle (silica co-polymer) stationary phase, providing the efficiency of silica with the stability of polymers. This C8 column is designed for Mass Spectrometry applications, offering sharp peaks, good sensitivity, and large peak capacities.
  • Designed with trifunctional bonding chemistries for long lifetimes over a wide pH range.
  • Provides high throughput, excellent resolution, and ultra-low bleed.
  • Features homogenous particle coverage.
  • Contains one-third less silanols for superior peak shape.
  • Operates within a pH range of 1–12.
  • Constructed from hybrid particle (silica co-polymer) material.
  • Utilizes methylene bridged particles with 12% carbon load.