Learn More
Waters Corp XTerra MS C8 VanGuard Cartridge, 125Å, 3.5 µm, 2.1 mm X 5 mm, 3/pk
Catalog No. NC1588417 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C8 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. XTerra MS C8 analytical columns containing 3.5 µm sorbent particles.
Catalog No. NC1588417 Supplier Waters Corp Supplier No. 186007905
XTerra MS C8 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. XTerra MS C8 analytical columns containing 3.5 µm sorbent particles.