Learn More
Waters Corp Waters XTerra MS C18 Intelligent Speed Column, 3.5 μm, 4.6 mm × 20 mm
Catalog No. 50780792 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 Intelligent Speed Columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications. They offer superior pH stability compared to silica-based columns, sharp peaks, good sensitivity, and large peak capacities. With trifunctional bonding chemistries, these columns provide long lifetimes over a wide pH range, high throughput, excellent resolution, and ultra-low bleed.
  • Hybrid-based, reversed-phase C18 chemistry.
  • Designed for mass spectrometry applications, with superior pH stability.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Features a wide pH range of 1 to 12 pH and maximum pressure of 6000 psi (415 Bar).
  • Spherical particle shape with 3.5 μm size and 125 Å pore size.
  • Dimensions: 4.6 mm inner diameter and 20 mm length.
  • Intelligent Speed (IS) format for HPLC systems.
Catalog No. 50-780-792 Supplier Waters Corp Supplier No. 186001891
XTerra MS C18 Intelligent Speed Columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications. They offer superior pH stability compared to silica-based columns, sharp peaks, good sensitivity, and large peak capacities. With trifunctional bonding chemistries, these columns provide long lifetimes over a wide pH range, high throughput, excellent resolution, and ultra-low bleed.
  • Hybrid-based, reversed-phase C18 chemistry.
  • Designed for mass spectrometry applications, with superior pH stability.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Features a wide pH range of 1 to 12 pH and maximum pressure of 6000 psi (415 Bar).
  • Spherical particle shape with 3.5 μm size and 125 Å pore size.
  • Dimensions: 4.6 mm inner diameter and 20 mm length.
  • Intelligent Speed (IS) format for HPLC systems.