Learn More
Waters Corp XTerra Phenyl Column, 125 Å, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp 186001147RF
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled device-tagged columns were developed to automatically identify, track, and digitally trace column use history. While eConnect columns can be used on any HPLC, their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • Contains unique column-specific information with the eConnect column tag.
  • Automatic identification, tracking, and digital tracing for column use history.
  • Compatible with any HPLC system.
  • Enhanced functionality is achieved when integrated with compatible intelligent systems and software.
Catalog No. 50-239-2516