Learn More
Waters Corp Waters XTerra HPLC Column, 3.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186001180
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first-generation columns provide superior pH stability compared to a silica-based column.
  • Superior pH stability compared to a silica-based column.
Catalog No. 50-780-611