Learn More
Waters Corp Waters Column, Phenyl, 125 Å, 3.5 μm, 4.6 mm x 250 mm
Catalog No. 50780703 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These hybrid-based, reversed-phase columns with phenyl ligand functionality provide superior pH stability over silica-based columns. They offer complementary selectivity for complex mixture separations, utilizing Hybrid Particle Technology that combines inorganic (silica) and organic (organosiloxane) components for high efficiency.
  • Hybrid-based, reversed-phase with phenyl ligand functionality.
  • Superior pH stability compared to silica-based columns.
  • Complementary selectivity for compounds with aromatic rings.
  • Excellent peak shape and reproducible retention times.
  • Difunctional bonding chemistry ensures improved low pH hydrolytic stability.
  • Manufactured with strict quality control and ultrapure reagents.
  • Includes certificates of batch analysis and column efficiency.
  • Wide operating pH range of 1–12.
Catalog No. 50-780-703 Supplier Waters Corp Supplier No. 186001474
These hybrid-based, reversed-phase columns with phenyl ligand functionality provide superior pH stability over silica-based columns. They offer complementary selectivity for complex mixture separations, utilizing Hybrid Particle Technology that combines inorganic (silica) and organic (organosiloxane) components for high efficiency.
  • Hybrid-based, reversed-phase with phenyl ligand functionality.
  • Superior pH stability compared to silica-based columns.
  • Complementary selectivity for compounds with aromatic rings.
  • Excellent peak shape and reproducible retention times.
  • Difunctional bonding chemistry ensures improved low pH hydrolytic stability.
  • Manufactured with strict quality control and ultrapure reagents.
  • Includes certificates of batch analysis and column efficiency.
  • Wide operating pH range of 1–12.