Learn More
Waters Corp Waters Column, Phenyl, 3.5 μm, 3.9 mm x 100 mm
Catalog No. 50780608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These hybrid-based, reversed-phase columns incorporate phenyl ligand functionality. They provide superior pH stability compared to silica-based columns.
  • Offers complementary selectivity.
  • Delivers excellent peak shapes for all compounds.
  • Operates within a pH range of 1 to 12.
  • Features difunctional bonding.
  • Ideally suited for mass spectrometry applications.
  • Utilizes first-generation hybrid particle technology.
  • Provides high mechanical strength and high efficiency.
  • Has a spherical particle shape.
  • Features a pore size of 125 Å.
  • Ensures homogeneous coverage due to bonded and endcapped particles.
  • Contains 1/3 less silanols compared to conventional silica-based materials, contributing to superior peak shape.
Catalog No. 50-780-608 Supplier Waters Corp Supplier No. 186001177
These hybrid-based, reversed-phase columns incorporate phenyl ligand functionality. They provide superior pH stability compared to silica-based columns.
  • Offers complementary selectivity.
  • Delivers excellent peak shapes for all compounds.
  • Operates within a pH range of 1 to 12.
  • Features difunctional bonding.
  • Ideally suited for mass spectrometry applications.
  • Utilizes first-generation hybrid particle technology.
  • Provides high mechanical strength and high efficiency.
  • Has a spherical particle shape.
  • Features a pore size of 125 Å.
  • Ensures homogeneous coverage due to bonded and endcapped particles.
  • Contains 1/3 less silanols compared to conventional silica-based materials, contributing to superior peak shape.