Learn More
Waters Corp Waters Corporation Column, Phenyl, 125 Å, 3.5 μm, 3 x 50 mm
Catalog No. 50780573 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first-generation columns provide superior pH stability compared to a silica-based column.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • 2-methyl-2-phenylethyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 125 Å pore size
  • For HPLC systems
  • 3 mm inner diameter
  • 50 mm length
  • 12% carbon load
  • 1 unit per package
Catalog No. 50-780-573 Supplier Waters Corp Supplier No. 186001141
Add to Cart
Add to Cart
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first-generation columns provide superior pH stability compared to a silica-based column.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • 2-methyl-2-phenylethyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 125 Å pore size
  • For HPLC systems
  • 3 mm inner diameter
  • 50 mm length
  • 12% carbon load
  • 1 unit per package