Learn More
Waters Corp Waters XTerra Phenyl Column, 125 Å, 3.5 μm, 3.9 mm × 150 mm
Catalog No. 50780609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XTerra Phenyl Column is a hybrid-based, reversed-phase column offering superior pH stability and unique selectivity for complex mixture separations, particularly for aromatic rings. It utilizes Hybrid Particle Technology for high separation efficiency and enhanced pH stability.
  • Enhanced pH stability (1 to 12 pH range) due to its hybrid particle substrate.
  • Provides unique selectivity for aromatic compounds and complex mixture separations.
  • Ensures reproducible retention and excellent peak shape; features low silanol activity.
  • Endcapped for improved performance, offering high efficiency and rugged mechanical strength.
  • Compatible with mass spectrometry and designed for long lifetimes.
  • Facilitates easy scale-up and allows for increased mass loading at high pH.
Catalog No. 50-780-609 Supplier Waters Corp Supplier No. 186001178
Add to Cart
Add to Cart
This XTerra Phenyl Column is a hybrid-based, reversed-phase column offering superior pH stability and unique selectivity for complex mixture separations, particularly for aromatic rings. It utilizes Hybrid Particle Technology for high separation efficiency and enhanced pH stability.
  • Enhanced pH stability (1 to 12 pH range) due to its hybrid particle substrate.
  • Provides unique selectivity for aromatic compounds and complex mixture separations.
  • Ensures reproducible retention and excellent peak shape; features low silanol activity.
  • Endcapped for improved performance, offering high efficiency and rugged mechanical strength.
  • Compatible with mass spectrometry and designed for long lifetimes.
  • Facilitates easy scale-up and allows for increased mass loading at high pH.