Learn More
Waters Corp XTerra Phenyl Column, 125Å, 3.5 µm, 4.6 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186001139
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-571