Learn More
Waters Corp Waters XTerra HPLC Column, 3.5 μm, Phenyl, 3.0 x 100 mm
Supplier:  Waters Corp 186001142
Encompass_Preferred
This HPLC column features a 3.5 μm particle size and a phenyl ligand type, with dimensions of 3.0 x 100 mm. It utilizes a hybrid particle stationary phase, providing ruggedness, high mechanical strength, and efficiency across a broad pH range.
  • Offers complementary selectivity to other phases
  • Delivers excellent peak shapes for all compounds
  • Features difunctional bonding
  • pH range of 1–12
  • Ideally suited for mass spectrometry applications
  • Particle provides homogeneous coverage and superior peak shape
  • One-third less silanols compared to conventional silica
Catalog No. 50-780-574