Learn More
Waters Corp XTerra Phenyl Column, 125Å, 3.5 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. 50818714 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-818-714 Supplier Waters Corp Supplier No. 186001140
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first generation columns provide superior pH stability compared to a silica-based column.