Learn More
Waters Corp Column, XTerra Phenyl, 5 μm, 4.6 mm id, 250 mm Length
Supplier:  Waters Corp 186001147
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns designed to incorporate phenyl ligand functionality. These first-generation columns offer superior pH stability compared to traditional silica-based columns.
  • Hybrid-based, reversed-phase columns with phenyl ligand functionality
  • Superior pH stability compared to silica-based columns
  • Spherical particle shape
  • 5 μm particle size
  • 125Å pore size
  • Low silanol activity
  • pH range of 1 to 12
  • 12% carbon load
  • 170 surface area
Catalog No. 50-780-579