Learn More
Waters Corp XTerra Phenyl VanGuard Cartridge, 125Å, 3.5 µm, 2.1 mm X 5 mm, 3/pk
Catalog No. NC1719819 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Phenyl VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. XTerra Phenyl analytical columns containing 3.5 µm sorbent particles.
Catalog No. NC1719819 Supplier Waters Corp Supplier No. 186007917
XTerra Phenyl VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. XTerra Phenyl analytical columns containing 3.5 µm sorbent particles.