Learn More
Waters Corp VanGuard Cartridge, XTerra Phenyl, 2.1 x 5 mm, 5 μm
Catalog No. 502392652 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is an XTerra Phenyl VanGuard Cartridge, which utilizes first-generation hybrid particle stationary phase technology. This technology combines the efficiency of silica with the stability of polymers, offering high mechanical strength and excellent peak shapes for various compounds over a wide pH range.
  • Complementary selectivity to other phases.
  • Delivers excellent peak shapes for all compounds.
  • Features difunctional bonding.
  • Ideally suited for Mass Spec applications.
  • Can be used with a universal cartridge holder.
  • Particle size: 5 μm.
  • Dimensions: 2.1 x 5 mm.
Catalog No. 50-239-2652 Supplier Waters Corp Supplier No. 186007921
This product is an XTerra Phenyl VanGuard Cartridge, which utilizes first-generation hybrid particle stationary phase technology. This technology combines the efficiency of silica with the stability of polymers, offering high mechanical strength and excellent peak shapes for various compounds over a wide pH range.
  • Complementary selectivity to other phases.
  • Delivers excellent peak shapes for all compounds.
  • Features difunctional bonding.
  • Ideally suited for Mass Spec applications.
  • Can be used with a universal cartridge holder.
  • Particle size: 5 μm.
  • Dimensions: 2.1 x 5 mm.