Learn More
Waters Corp WATERS CORPORATION Prep Guard Cartridge, C18, Hybrid, 10 mm x 10 mm
Catalog No. 50780526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns utilize patented Hybrid Particle Technology, specifically engineered for preparative isolation and purification in discovery and development laboratories. They operate routinely at pH 1–12, enabling up to 60X greater loading capacity.
  • Incorporates patented Hybrid Particle Technology for preparative applications.
  • Operates routinely at pH 1–12.
  • Offers up to 60X greater loading capacity for fewer injections and faster results.
  • Optimized for mass spectrometry applications.
  • Delivers sharp peaks, good sensitivity, and large peak capacities.
  • Features trifunctional bonding chemistries for extended lifetimes.
  • Provides high throughput, excellent resolution, and ultra-low bleed.
  • Has a 125 Å pore size.
  • Operates within temperature limits of 20°C to 60°C.
Catalog No. 50-780-526 Supplier Waters Corp Supplier No. 186001001
These columns utilize patented Hybrid Particle Technology, specifically engineered for preparative isolation and purification in discovery and development laboratories. They operate routinely at pH 1–12, enabling up to 60X greater loading capacity.
  • Incorporates patented Hybrid Particle Technology for preparative applications.
  • Operates routinely at pH 1–12.
  • Offers up to 60X greater loading capacity for fewer injections and faster results.
  • Optimized for mass spectrometry applications.
  • Delivers sharp peaks, good sensitivity, and large peak capacities.
  • Features trifunctional bonding chemistries for extended lifetimes.
  • Provides high throughput, excellent resolution, and ultra-low bleed.
  • Has a 125 Å pore size.
  • Operates within temperature limits of 20°C to 60°C.