Learn More
Waters Corp HPLC Guard Cartridge, XTerra MS C18, 5 μm, 19 x 10 mm
Catalog No. 50780562 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparatory guard cartridge is engineered with Hybrid Particle Technology to deliver enhanced separation performance and column longevity. It features a unique silica/polymer hybrid, which provides superior pH stability compared to traditional silica-based columns, allowing for extended column lifetimes and robust method development. This cartridge is ideal for demanding preparative chromatography applications where high efficiency and durability are crucial.
  • Utilizes a unique silica/polymer hybrid for enhanced performance.
  • Offers superior pH stability, broadening the range of compatible mobile phases.
  • Designed for demanding preparative chromatography.
  • Contributes to extended column lifetimes.
  • Ensures robust method development.
Catalog No. 50-780-562 Supplier Waters Corp Supplier No. 186001104
This preparatory guard cartridge is engineered with Hybrid Particle Technology to deliver enhanced separation performance and column longevity. It features a unique silica/polymer hybrid, which provides superior pH stability compared to traditional silica-based columns, allowing for extended column lifetimes and robust method development. This cartridge is ideal for demanding preparative chromatography applications where high efficiency and durability are crucial.
  • Utilizes a unique silica/polymer hybrid for enhanced performance.
  • Offers superior pH stability, broadening the range of compatible mobile phases.
  • Designed for demanding preparative chromatography.
  • Contributes to extended column lifetimes.
  • Ensures robust method development.