Learn More
Waters Corp Waters Guard Cartridge, XTerra MS C18, 7.8 x 10 mm
Catalog No. 50780600 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra MS C18 Guard Cartridge is a 7.8 x 10 mm guard cartridge (5 μm particle, 125 Å pore size) engineered for Mass Spectrometry. It uses hybrid particle technology for silica efficiency, polymer stability, and an expanded pH range (1–12).
  • Hybrid particle combines silica efficiency with polymer stability.
  • Offers high mechanical strength, efficiency, and excellent peak shape for bases.
  • Wider pH range (1–12) than silica-based particles.
  • Engineered for Mass Spectrometry: sharp peaks, good sensitivity, large peak capacities.
  • Trifunctional bonding chemistries: long lifetimes, high throughput, resolution, ultra-low bleed.
  • Facilitates analytical to preparative chromatography scale-up.
  • High pH operation for excellent peak shapes and increased mass loading (up to 60×).
Catalog No. 50-780-600 Supplier Waters Corp Supplier No. 186001168
The XTerra MS C18 Guard Cartridge is a 7.8 x 10 mm guard cartridge (5 μm particle, 125 Å pore size) engineered for Mass Spectrometry. It uses hybrid particle technology for silica efficiency, polymer stability, and an expanded pH range (1–12).
  • Hybrid particle combines silica efficiency with polymer stability.
  • Offers high mechanical strength, efficiency, and excellent peak shape for bases.
  • Wider pH range (1–12) than silica-based particles.
  • Engineered for Mass Spectrometry: sharp peaks, good sensitivity, large peak capacities.
  • Trifunctional bonding chemistries: long lifetimes, high throughput, resolution, ultra-low bleed.
  • Facilitates analytical to preparative chromatography scale-up.
  • High pH operation for excellent peak shapes and increased mass loading (up to 60×).