Learn More
Waters Corp XTerra MS C18 Prep Guard Cartridge, 125Å, 10 μm
Catalog No. 50780527 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Prep columns incorporate Hybrid Particle Technology for isolation and purification applications in discovery and development labs. Engineered for preparative use, they allow routine operation at pH 1 to 12, enabling up to 60 times more loading onto the column. This increases capacity, reducing injections and providing more material. Shorter columns can produce the same amount of material in half the time. This guard cartridge protects prep columns from contaminants and extends column lifetimes; a separate cartridge holder is required for proper installation.
  • Utilizes hybrid particle technology for isolation and purification.
  • Engineered for preparative applications.
  • Operates at pH 1 to 12.
  • Enables up to 60X more column loading.
  • Protects prep columns from contaminants.
  • Extends column lifetimes.
  • Requires a cartridge holder for installation.
Catalog No. 50-780-527 Supplier Waters Corp Supplier No. 186001002
XTerra Prep columns incorporate Hybrid Particle Technology for isolation and purification applications in discovery and development labs. Engineered for preparative use, they allow routine operation at pH 1 to 12, enabling up to 60 times more loading onto the column. This increases capacity, reducing injections and providing more material. Shorter columns can produce the same amount of material in half the time. This guard cartridge protects prep columns from contaminants and extends column lifetimes; a separate cartridge holder is required for proper installation.
  • Utilizes hybrid particle technology for isolation and purification.
  • Engineered for preparative applications.
  • Operates at pH 1 to 12.
  • Enables up to 60X more column loading.
  • Protects prep columns from contaminants.
  • Extends column lifetimes.
  • Requires a cartridge holder for installation.