Learn More
Waters Corp Waters Prep Column, 125Å, 10μm, 7.8 mm × 300 mm
Catalog No. 50780596 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Prep columns utilize Hybrid Particle Technology for isolation and purification in discovery and development laboratories. Patented Hybrid Particle Technology for prep enables routine operation at pH 1 to 12. This allows loading up to 60 times more onto the column, resulting in fewer injections or more material. Greater capacity also means a shorter column can produce the same material in half the time.
  • Hybrid Particle Technology for isolation and purification.
  • Patented Hybrid Particle Technology engineered for prep; routine operation at pH 1 to 12.
  • Load up to 60 times more onto the column, reducing injections or increasing material yield.
  • Tough material with outstanding mechanical strength and efficiency.
  • Excellent peak shapes for bases; simple scale-up from analytical to preparative chromatography.
  • Excellent chemical stability for low and high pH purifications.
Catalog No. 50-780-596 Supplier Waters Corp Supplier No. 186001164
XTerra Prep columns utilize Hybrid Particle Technology for isolation and purification in discovery and development laboratories. Patented Hybrid Particle Technology for prep enables routine operation at pH 1 to 12. This allows loading up to 60 times more onto the column, resulting in fewer injections or more material. Greater capacity also means a shorter column can produce the same material in half the time.
  • Hybrid Particle Technology for isolation and purification.
  • Patented Hybrid Particle Technology engineered for prep; routine operation at pH 1 to 12.
  • Load up to 60 times more onto the column, reducing injections or increasing material yield.
  • Tough material with outstanding mechanical strength and efficiency.
  • Excellent peak shapes for bases; simple scale-up from analytical to preparative chromatography.
  • Excellent chemical stability for low and high pH purifications.