Learn More
Waters Corp Waters PREPARATIVE COLUMN, 10 μm, 50 × 150 mm, MS C18
Catalog No. 50781396 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XTerra MS C18 PREPARATIVE COLUMN is designed for mass spectrometry applications, delivering sharp peaks, high sensitivity, and large peak capacities. Its trifunctional bonding chemistry ensures a long lifespan across a wide pH range, providing excellent resolution and ultra-low bleed for high-throughput analysis.
  • Delivers sharp peaks, high sensitivity, and large peak capacities.
  • Features trifunctional bonding for long column lifetimes over a pH range of 1 to 12.
  • Provides high throughput with excellent resolution and ultra-low bleed.
  • Spherical particles: 125 Å pore, 15.5% carbon load, end-capped.
  • Operating temperature range: 20°C to 60°C.
  • Hybrid particle design offers high mechanical strength.
  • Enables easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60×).
Catalog No. 50-781-396 Supplier Waters Corp Supplier No. 186002843
Add to Cart
Add to Cart
Request a Quote
This XTerra MS C18 PREPARATIVE COLUMN is designed for mass spectrometry applications, delivering sharp peaks, high sensitivity, and large peak capacities. Its trifunctional bonding chemistry ensures a long lifespan across a wide pH range, providing excellent resolution and ultra-low bleed for high-throughput analysis.
  • Delivers sharp peaks, high sensitivity, and large peak capacities.
  • Features trifunctional bonding for long column lifetimes over a pH range of 1 to 12.
  • Provides high throughput with excellent resolution and ultra-low bleed.
  • Spherical particles: 125 Å pore, 15.5% carbon load, end-capped.
  • Operating temperature range: 20°C to 60°C.
  • Hybrid particle design offers high mechanical strength.
  • Enables easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60×).