Learn More
Waters Corp WATERS CORPORATION XTerra Prep MS C18 Column, 10 μm, 30 x 300 mm
Supplier:  Waters Corp 186002275
Encompass_Preferred
This XTerra Prep MS C18 Column is designed for Mass Spectrometry applications, providing sharp peaks, good sensitivity, and large peak capacities. Its trifunctional bonding chemistries ensure long lifetimes over a wide pH range of 1 to 12, offering high throughput with excellent resolution and ultra-low bleed.
  • Compatible with Mass Spectrometry applications.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Delivers long lifetimes over a wide pH range of 1 to 12.
  • Offers high throughput, excellent resolution, and ultra-low bleed.
  • Features homogenous coverage, 1/3 less silanols, and superior peak shape.
  • Constructed from a rugged material with high mechanical strength.
  • Ensures excellent peak shape for bases and increased mass loading at high pH.
  • Allows for easy scale-up from analytical to preparative chromatography.
Catalog No. 50-781-051
Add to Cart
Request a Quote