Learn More
Waters Corp Waters Preparative Column, C18, 5 μm, 50 mm ID, 100 mm Length
Catalog No. 50781023 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Prep columns incorporate Hybrid Particle Technology for isolation and purification in discovery and development laboratories. XTerra Prep MS columns feature patented Hybrid Particle Technology, with particles specifically engineered for preparative applications. This technology allows for operation at pH 1–12, enabling up to 60X more loading capacity onto the column. This increased capacity results in fewer injections to obtain the same amount of pure material or more material with the same number of injections. For discovery applications, this greater capacity allows for the use of a shorter column, producing the same amount of material in half the time.
  • Patented Hybrid Particle Technology.
  • Operates routinely at pH 1–12.
  • Load up to 60X more onto the column.
  • Allows for use of shorter columns to achieve the same material in half the time in discovery.
Catalog No. 50-781-023 Supplier Waters Corp Supplier No. 186002222
Add to Cart
Add to Cart
Request a Quote
XTerra Prep columns incorporate Hybrid Particle Technology for isolation and purification in discovery and development laboratories. XTerra Prep MS columns feature patented Hybrid Particle Technology, with particles specifically engineered for preparative applications. This technology allows for operation at pH 1–12, enabling up to 60X more loading capacity onto the column. This increased capacity results in fewer injections to obtain the same amount of pure material or more material with the same number of injections. For discovery applications, this greater capacity allows for the use of a shorter column, producing the same amount of material in half the time.
  • Patented Hybrid Particle Technology.
  • Operates routinely at pH 1–12.
  • Load up to 60X more onto the column.
  • Allows for use of shorter columns to achieve the same material in half the time in discovery.