Learn More
Waters Corp Waters Corporation Guard Cartridge, XTerra MS C8, 19 x 10 mm, 10 μm, 125 Å
Supplier:  Waters Corp 186001035
Encompass_Preferred
This guard cartridge is designed for HPLC columns and is compatible with Mass Spectrometry applications. It features an XTerra MS C8 hybrid particle stationary phase, combining the efficiency of silica with an expanded pH range, making it suitable for a variety of chromatographic needs.
  • Designed for mass spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • High throughput with excellent resolution and ultra-low bleed
  • Enables easy scale-up from analytical to preparative chromatography
  • Operates at high pH for excellent peak shapes for bases and increased mass loading (up to 60x)
  • Particle features homogenous coverage and reduced silanols
  • Supports a pH range of 1–12
Catalog No. 50-780-558