Learn More
Waters Corp Waters Corporation XTerra MS C8 Guard Cartridge, 10 μm, 10 mm x 7.8 mm
Catalog No. 50780605 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra columns offer high efficiency separations, combining silica and polymer particle benefits for a rugged material with high mechanical strength and an expanded pH range. Designed for mass spectrometry, XTerra MS C8 columns deliver sharp peaks, good sensitivity, and large peak capacities. Trifunctional bonding chemistries ensure long lifetimes over a wide pH range, high throughput, excellent resolution, and ultra-low bleed.
  • Compatible with mass spectrometry applications
  • Sharp peaks, good sensitivity, large peak capacities
  • Trifunctional bonding chemistries for long lifetimes over a wide pH range
  • High throughput with excellent resolution and ultra-low bleed
  • Spherical particle shape
  • Pore size: 125 Å
  • Temperature limits: 20°C to 60°C
  • pH range: 1 to 12
Catalog No. 50-780-605 Supplier Waters Corp Supplier No. 186001173
XTerra columns offer high efficiency separations, combining silica and polymer particle benefits for a rugged material with high mechanical strength and an expanded pH range. Designed for mass spectrometry, XTerra MS C8 columns deliver sharp peaks, good sensitivity, and large peak capacities. Trifunctional bonding chemistries ensure long lifetimes over a wide pH range, high throughput, excellent resolution, and ultra-low bleed.
  • Compatible with mass spectrometry applications
  • Sharp peaks, good sensitivity, large peak capacities
  • Trifunctional bonding chemistries for long lifetimes over a wide pH range
  • High throughput with excellent resolution and ultra-low bleed
  • Spherical particle shape
  • Pore size: 125 Å
  • Temperature limits: 20°C to 60°C
  • pH range: 1 to 12