Learn More
Waters Corp Waters XTerra MS C8 Prep Guard Cartridge, 125Å, 5 μm, 19 mm x 10 mm
Catalog No. 50780563 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XTerra MS C8 Prep Guard Cartridge delivers high-efficiency separations in mass spectrometry applications. It features a hybrid particle stationary phase, combining silica's efficiency with polymer stability for robust performance.
  • Provides an expanded pH range of a polymer particle.
  • Features a rugged material with high mechanical strength.
  • Delivers excellent peak shape for bases.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Utilizes trifunctional bonding chemistries for long lifetimes over a wide pH range.
  • Enables high throughput with excellent resolution and ultra-low bleed.
  • Allows for easy scale-up from analytical to preparative chromatography.
  • Operates at high pH, offering excellent peak shapes for bases and increased mass loading.
Catalog No. 50-780-563 Supplier Waters Corp Supplier No. 186001105
This XTerra MS C8 Prep Guard Cartridge delivers high-efficiency separations in mass spectrometry applications. It features a hybrid particle stationary phase, combining silica's efficiency with polymer stability for robust performance.
  • Provides an expanded pH range of a polymer particle.
  • Features a rugged material with high mechanical strength.
  • Delivers excellent peak shape for bases.
  • Provides sharp peaks, good sensitivity, and large peak capacities.
  • Utilizes trifunctional bonding chemistries for long lifetimes over a wide pH range.
  • Enables high throughput with excellent resolution and ultra-low bleed.
  • Allows for easy scale-up from analytical to preparative chromatography.
  • Operates at high pH, offering excellent peak shapes for bases and increased mass loading.