Learn More
Waters Corp Waters Prep Column, C8, 125Å, 5 μm, 19 mm x 100 mm
Supplier:  Waters Corp 186001935
Encompass_Preferred
XTerra Prep columns utilize Hybrid Particle Technology for isolation and purification. Engineered for preparative applications, they operate at pH 1 to 12, offering up to 60X more loading capacity, which reduces injections or yields more material.
  • Hybrid particle technology for isolation and purification
  • Engineered for preparative applications
  • Operates routinely at pH 1 to 12
  • Up to 60X more loading capacity
  • C8 chemistry
  • 5 μm particle size
  • 125 Å pore size
  • 19 mm inner diameter x 100 mm length
  • Maximum pressure 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • Suitable for LC/MS and LC techniques
Catalog No. 50-780-826