Learn More
Waters Corp Waters HPLC Column, MS C8, 5 μm, 7.8 x 50 mm
Catalog No. 50780585 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HPLC columns feature first-generation hybrid particle (silica co-polymer) stationary phases with methylene-bridged particles. Designed for Mass Spectrometry applications, they provide sharp peaks, good sensitivity, and large peak capacities.
  • Trifunctional bonding chemistries ensure long lifetimes across a wide pH range (pH 1–12).
  • Achieves high throughput with excellent resolution and ultra-low bleed.
  • Combines high efficiency separations of silica with the expanded pH range of a polymer.
  • Rugged material with high mechanical strength and excellent peak shape for bases.
  • Homogenous coverage and 1/3 less silanols contribute to superior peak shape.
  • Suitable for preparative chromatography, allowing scale-up from analytical methods.
  • Increased mass loading up to 60x compared to conventional silica-based materials.
Catalog No. 50-780-585 Supplier Waters Corp Supplier No. 186001153
These HPLC columns feature first-generation hybrid particle (silica co-polymer) stationary phases with methylene-bridged particles. Designed for Mass Spectrometry applications, they provide sharp peaks, good sensitivity, and large peak capacities.
  • Trifunctional bonding chemistries ensure long lifetimes across a wide pH range (pH 1–12).
  • Achieves high throughput with excellent resolution and ultra-low bleed.
  • Combines high efficiency separations of silica with the expanded pH range of a polymer.
  • Rugged material with high mechanical strength and excellent peak shape for bases.
  • Homogenous coverage and 1/3 less silanols contribute to superior peak shape.
  • Suitable for preparative chromatography, allowing scale-up from analytical methods.
  • Increased mass loading up to 60x compared to conventional silica-based materials.