Learn More
Waters Corp Waters Prep MS C8 Column, 5 μm, 19 x 150 mm, Hybrid Particle
Catalog No. 50781088 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column is designed for mass spectrometry applications. It features a 5 μm particle size, 19 x 150 mm dimensions, and hybrid particle technology, offering a wide pH range and excellent peak performance.
  • Compatible with Mass Spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Silica separations at polymer pH, combining high efficiency with expanded pH range
  • Rugged material with high mechanical strength and excellent peak shape for bases
  • Wider pH range (1–12) than silica–based particles
  • Homogenous coverage
  • 1/3 less silanols
  • Superior peak shape
Catalog No. 50-781-088 Supplier Waters Corp Supplier No. 186002380
This preparative column is designed for mass spectrometry applications. It features a 5 μm particle size, 19 x 150 mm dimensions, and hybrid particle technology, offering a wide pH range and excellent peak performance.
  • Compatible with Mass Spectrometry applications
  • Provides sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range
  • Provides high throughput with excellent resolution and ultra–low bleed
  • Silica separations at polymer pH, combining high efficiency with expanded pH range
  • Rugged material with high mechanical strength and excellent peak shape for bases
  • Wider pH range (1–12) than silica–based particles
  • Homogenous coverage
  • 1/3 less silanols
  • Superior peak shape