Learn More
Waters Corp WATERS CORPORATION XTerra Prep MS C8 Column, 5 μm Particle Size, 50 mm x 100 mm, 125 Å Pore Size
Catalog No. 50781024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Hybrid particle (silica co-polymer) technology provides efficiency and stability.
  • Wide pH range (1–12) for enhanced selectivity and improved peak shape.
  • Rugged material offers high mechanical strength and high efficiency.
  • Provides excellent peak shape for bases.
  • Designed for mass spectrometry (MS) applications, delivering sharp peaks and good sensitivity.
  • Trifunctional bonding chemistries ensure long lifetime and ultra-low bleed.
  • Easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60x) at high pH.
  • Carbon load: 12%.
  • Proprietary end-capping.
  • Operates within a temperature range of 20°C to 60°C.
  • USP Class No. L7.
Catalog No. 50-781-024 Supplier Waters Corp Supplier No. 186002223
Add to Cart
Add to Cart
Request a Quote
  • Hybrid particle (silica co-polymer) technology provides efficiency and stability.
  • Wide pH range (1–12) for enhanced selectivity and improved peak shape.
  • Rugged material offers high mechanical strength and high efficiency.
  • Provides excellent peak shape for bases.
  • Designed for mass spectrometry (MS) applications, delivering sharp peaks and good sensitivity.
  • Trifunctional bonding chemistries ensure long lifetime and ultra-low bleed.
  • Easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60x) at high pH.
  • Carbon load: 12%.
  • Proprietary end-capping.
  • Operates within a temperature range of 20°C to 60°C.
  • USP Class No. L7.