Learn More
Waters Corp Waters OBD Prep Column, C8, 5 μm, 50 mm x 50 mm
Catalog No. 50781020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Prep columns utilize patented Hybrid Particle Technology, specifically designed for preparative applications in discovery and development labs. They offer routine operation at pH 1 to 12, with up to 60 times more loading capacity. This increased capacity reduces injections for pure material and enables shorter columns for faster results.
  • Hybrid Particle Technology for isolation and purification.
  • Patented design specifically for preparative applications.
  • Wide pH range (1 to 12) for broad operational flexibility.
  • High loading capacity, up to 60 times more.
  • Increased efficiency: fewer injections or more material per injection.
  • Time savings in discovery through shorter columns for faster material production.
Catalog No. 50-781-020 Supplier Waters Corp Supplier No. 186002219
Add to Cart
Add to Cart
Request a Quote
XTerra Prep columns utilize patented Hybrid Particle Technology, specifically designed for preparative applications in discovery and development labs. They offer routine operation at pH 1 to 12, with up to 60 times more loading capacity. This increased capacity reduces injections for pure material and enables shorter columns for faster results.
  • Hybrid Particle Technology for isolation and purification.
  • Patented design specifically for preparative applications.
  • Wide pH range (1 to 12) for broad operational flexibility.
  • High loading capacity, up to 60 times more.
  • Increased efficiency: fewer injections or more material per injection.
  • Time savings in discovery through shorter columns for faster material production.