Learn More
Waters Corp Waters Corporation Prep MS C18 Column, 5 μm, 125 Å, 30 x 50 mm
Catalog No. 50818627 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This prep MS C18 column is designed for high-efficiency separations in mass spectrometry applications. It features a 5 μm particle size and 125 Å pore size, offering ruggedness and an expanded pH range for versatile use.
  • High efficiency separations
  • Expanded pH range (1–12)
  • Rugged material with high mechanical strength
  • Excellent peak shape for bases
  • Superior peak shape due to homogenous coverage and 1/3 less silanols
  • Compatible with Mass Spectrometry applications
  • Delivers sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries provide long lifetimes over a wide pH range
  • High throughput with excellent resolution and ultra-low bleed
  • Easy scale-up from analytical to preparative chromatography
  • Allows operation at high pH for increased mass loading compared to conventional silica-based materials.
Catalog No. 50-818-627 Supplier Waters Corp Supplier No. 186001938
This prep MS C18 column is designed for high-efficiency separations in mass spectrometry applications. It features a 5 μm particle size and 125 Å pore size, offering ruggedness and an expanded pH range for versatile use.
  • High efficiency separations
  • Expanded pH range (1–12)
  • Rugged material with high mechanical strength
  • Excellent peak shape for bases
  • Superior peak shape due to homogenous coverage and 1/3 less silanols
  • Compatible with Mass Spectrometry applications
  • Delivers sharp peaks, good sensitivity, and large peak capacities
  • Trifunctional bonding chemistries provide long lifetimes over a wide pH range
  • High throughput with excellent resolution and ultra-low bleed
  • Easy scale-up from analytical to preparative chromatography
  • Allows operation at high pH for increased mass loading compared to conventional silica-based materials.